Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011915.hg.1 |
TC0X000891.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011915.hg.1 |
ENST00000331511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011915.hg.1 |
ENST00000360011.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011915.hg.1 |
ENST00000451717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011915.hg.1 |
ENST00000545871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011915.hg.1 |
NM_001172739.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011915.hg.1 |
NM_001172743.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011915.hg.1 |
NM_021785.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011915.hg.1 |
NR_033348.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011915.hg.1 |
NR_033349.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011914.hg.1 |
TC0X000891.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011914.hg.1 |
ENST00000331511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011914.hg.1 |
ENST00000360011.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011914.hg.1 |
ENST00000451717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011914.hg.1 |
ENST00000509491.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011914.hg.1 |
ENST00000545871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011914.hg.1 |
NM_001172739.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011914.hg.1 |
NM_001172743.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011914.hg.1 |
NM_021785.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011914.hg.1 |
NR_033348.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011914.hg.1 |
NR_033349.1 |
Refseq |