Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021120.hg.1 |
TC04001488.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021120.hg.1 |
ENST00000264363.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021120.hg.1 |
NM_022569.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000264363.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000264363.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000264363.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000264363.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000264363.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000264363.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000264363.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000264363.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000264363.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000264363.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000264363.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000264363.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000264363.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000264363.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000264363.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000264363.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000264363.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000264363.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000504854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000504854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000504854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000504854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000504854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000504854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000504854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000504854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000504854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000504854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000504854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000504854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000504854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000504854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000514570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000514570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000514570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000514570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000514570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000514570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000514570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000514570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
NM_022569.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
NM_022569.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
NM_022569.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
NM_022569.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
NM_022569.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
NM_022569.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
NM_022569.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
NM_022569.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
NM_022569.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
NM_022569.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
NM_022569.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
NM_022569.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
NM_022569.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
NM_022569.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
NM_022569.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
NM_022569.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
NM_022569.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011659.hg.1 |
NM_022569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000264363.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000264363.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000264363.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000264363.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000264363.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000264363.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000264363.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000264363.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000264363.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000264363.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000264363.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000264363.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000264363.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000264363.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000264363.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000264363.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000264363.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000264363.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000504854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000504854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000504854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000504854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000504854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000504854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000504854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000504854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000504854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000504854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000504854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000504854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000504854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000504854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000514570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000514570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000514570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000514570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000514570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000514570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000514570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000514570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
NM_022569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
NM_022569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
NM_022569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
NM_022569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
NM_022569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
NM_022569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
NM_022569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
NM_022569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
NM_022569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
NM_022569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
NM_022569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
NM_022569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
NM_022569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
NM_022569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
NM_022569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
NM_022569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
NM_022569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011659.hg.1 |
NM_022569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180074.hg.1 |
ENST00000264363.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180074.hg.1 |
ENST00000504854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180074.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180074.hg.1 |
NM_022569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180074.hg.1 |
ENST00000264363.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180074.hg.1 |
ENST00000504854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180074.hg.1 |
ENST00000613194.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180074.hg.1 |
NM_022569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180074.hg.1 |
TC0400011659.hg.1 |
Clariom D TC |