Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01049204.hg.1 |
TC01003208.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01049204.hg.1 |
ENST00000271732.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01049204.hg.1 |
ENST00000369003.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01049204.hg.1 |
ENST00000540514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01049204.hg.1 |
NM_018178.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015747.hg.1 |
ENST00000271732.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015747.hg.1 |
ENST00000271732.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015747.hg.1 |
ENST00000271732.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015747.hg.1 |
ENST00000271732.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015747.hg.1 |
ENST00000271732.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015747.hg.1 |
ENST00000271732.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015747.hg.1 |
ENST00000271732.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015747.hg.1 |
ENST00000427665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015747.hg.1 |
ENST00000427665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015747.hg.1 |
ENST00000427665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015747.hg.1 |
ENST00000427665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015747.hg.1 |
ENST00000427665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015747.hg.1 |
ENST00000427665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015747.hg.1 |
ENST00000427665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015747.hg.1 |
ENST00000427665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015747.hg.1 |
ENST00000479757.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015747.hg.1 |
ENST00000479757.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015747.hg.1 |
ENST00000479757.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015747.hg.1 |
ENST00000479757.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015747.hg.1 |
NM_018178.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015747.hg.1 |
NM_018178.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015747.hg.1 |
NM_018178.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015747.hg.1 |
NM_018178.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015747.hg.1 |
NM_018178.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015747.hg.1 |
NM_018178.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015747.hg.1 |
NM_018178.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015747.hg.1 |
NM_018178.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015747.hg.1 |
NM_018178.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015747.hg.1 |
NM_018178.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100217944.hg.1 |
ENST00000271732.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100217944.hg.1 |
ENST00000427665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100217944.hg.1 |
NM_018178.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100217944.hg.1 |
ENST00000271732.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100217944.hg.1 |
ENST00000427665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100217944.hg.1 |
NM_018178.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100217944.hg.1 |
TC0100015747.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100015747.hg.1 |
ENST00000271732.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015747.hg.1 |
ENST00000271732.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015747.hg.1 |
ENST00000271732.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015747.hg.1 |
ENST00000271732.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015747.hg.1 |
ENST00000271732.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015747.hg.1 |
ENST00000271732.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015747.hg.1 |
ENST00000271732.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015747.hg.1 |
ENST00000427665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015747.hg.1 |
ENST00000427665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015747.hg.1 |
ENST00000427665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015747.hg.1 |
ENST00000427665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015747.hg.1 |
ENST00000427665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015747.hg.1 |
ENST00000427665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015747.hg.1 |
ENST00000427665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015747.hg.1 |
ENST00000427665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015747.hg.1 |
ENST00000479757.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015747.hg.1 |
ENST00000479757.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015747.hg.1 |
ENST00000479757.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015747.hg.1 |
ENST00000479757.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015747.hg.1 |
NM_018178.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015747.hg.1 |
NM_018178.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015747.hg.1 |
NM_018178.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015747.hg.1 |
NM_018178.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015747.hg.1 |
NM_018178.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015747.hg.1 |
NM_018178.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015747.hg.1 |
NM_018178.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015747.hg.1 |
NM_018178.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015747.hg.1 |
NM_018178.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015747.hg.1 |
NM_018178.5 |
Refseq |