Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006741.hg.1 |
ENST00000380607.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006741.hg.1 |
ENST00000380607.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006741.hg.1 |
ENST00000380607.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006741.hg.1 |
ENST00000380607.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006741.hg.1 |
ENST00000380607.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006741.hg.1 |
ENST00000380607.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006741.hg.1 |
ENST00000380607.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006741.hg.1 |
ENST00000380607.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006741.hg.1 |
ENST00000380607.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006741.hg.1 |
ENST00000380607.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006741.hg.1 |
ENST00000380607.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006741.hg.1 |
ENST00000380607.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006741.hg.1 |
ENST00000467085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006741.hg.1 |
ENST00000467085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006741.hg.1 |
ENST00000467085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006741.hg.1 |
ENST00000467085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006741.hg.1 |
NM_003026.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006741.hg.1 |
NM_003026.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006741.hg.1 |
NM_003026.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006741.hg.1 |
NM_003026.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006741.hg.1 |
NM_003026.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006741.hg.1 |
NM_003026.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006741.hg.1 |
NM_003026.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006741.hg.1 |
NM_003026.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006741.hg.1 |
NM_003026.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006741.hg.1 |
NM_003026.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006741.hg.1 |
NM_003026.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006741.hg.1 |
NM_003026.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006741.hg.1 |
NM_003026.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006741.hg.1 |
NM_003026.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001011.hg.1 |
TC09000085.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001011.hg.1 |
ENST00000380607.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001011.hg.1 |
ENST00000397481.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001011.hg.1 |
ENST00000537391.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001011.hg.1 |
ENST00000541215.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001011.hg.1 |
NM_003026.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900145959.hg.1 |
ENST00000380607.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900145959.hg.1 |
NM_003026.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900145959.hg.1 |
ENST00000380607.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900145959.hg.1 |
NM_003026.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900145959.hg.1 |
TC0900006741.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900006741.hg.1 |
ENST00000380607.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006741.hg.1 |
ENST00000380607.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006741.hg.1 |
ENST00000380607.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006741.hg.1 |
ENST00000380607.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006741.hg.1 |
ENST00000380607.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006741.hg.1 |
ENST00000380607.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006741.hg.1 |
ENST00000380607.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006741.hg.1 |
ENST00000380607.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006741.hg.1 |
ENST00000380607.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006741.hg.1 |
ENST00000380607.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006741.hg.1 |
ENST00000380607.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006741.hg.1 |
ENST00000380607.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006741.hg.1 |
ENST00000467085.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006741.hg.1 |
ENST00000467085.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006741.hg.1 |
ENST00000467085.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006741.hg.1 |
ENST00000467085.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006741.hg.1 |
NM_003026.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006741.hg.1 |
NM_003026.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006741.hg.1 |
NM_003026.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006741.hg.1 |
NM_003026.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006741.hg.1 |
NM_003026.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006741.hg.1 |
NM_003026.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006741.hg.1 |
NM_003026.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006741.hg.1 |
NM_003026.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006741.hg.1 |
NM_003026.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006741.hg.1 |
NM_003026.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006741.hg.1 |
NM_003026.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006741.hg.1 |
NM_003026.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006741.hg.1 |
NM_003026.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900006741.hg.1 |
NM_003026.3 |
Refseq |