Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STOML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256673.1 | NP_001243602.1 | 2 - 3 | 363 | 53 | |
NM_001256674.1 | NP_001243603.1 | 2 - 3 | 363 | 53 | ||
GenBank mRNA | AF111800.1 | - | 2 - 3 | 312 | 53 | |
AK302051.1 | - | 2 - 3 | 318 | 53 | ||
AL109664.1 | - | 2 - 3 | 193 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 Homo sapiens Probe spans exons |