Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007298.hg.1 |
TC07000519.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007298.hg.1 |
ENST00000361669.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007298.hg.1 |
ENST00000546348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007298.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007300.hg.1 |
TC07000519.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007300.hg.1 |
ENST00000361669.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007300.hg.1 |
ENST00000536043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007300.hg.1 |
ENST00000546348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007300.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007299.hg.1 |
TC07000519.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007299.hg.1 |
ENST00000361669.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007299.hg.1 |
ENST00000536043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007299.hg.1 |
ENST00000546348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007299.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007305.hg.1 |
TC07000519.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007305.hg.1 |
ENST00000361669.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007305.hg.1 |
ENST00000394720.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007305.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007305.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007296.hg.1 |
TC07000519.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007296.hg.1 |
ENST00000361669.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007296.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007297.hg.1 |
TC07000519.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007297.hg.1 |
ENST00000361669.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007297.hg.1 |
ENST00000546348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007297.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007302.hg.1 |
TC07000519.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007302.hg.1 |
ENST00000361669.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007302.hg.1 |
ENST00000394720.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007302.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007302.hg.1 |
ENST00000536043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007302.hg.1 |
ENST00000546348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007302.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000421579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000421579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000421579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000421579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000441140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000441140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000441140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000454217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000454217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000454217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000454217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157153.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157153.hg.1 |
ENST00000454217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157153.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157153.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157153.hg.1 |
ENST00000454217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157153.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157153.hg.1 |
TC0700008240.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700157159.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157159.hg.1 |
ENST00000421579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157159.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157159.hg.1 |
ENST00000441140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157159.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157159.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157159.hg.1 |
ENST00000421579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157159.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157159.hg.1 |
ENST00000441140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157159.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157159.hg.1 |
TC0700008240.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700157160.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157160.hg.1 |
ENST00000421579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157160.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157160.hg.1 |
ENST00000441140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157160.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157160.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157160.hg.1 |
ENST00000421579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157160.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157160.hg.1 |
ENST00000441140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157160.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157160.hg.1 |
TC0700008240.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700157154.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157154.hg.1 |
ENST00000454217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157154.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157154.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157154.hg.1 |
ENST00000454217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157154.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157154.hg.1 |
TC0700008240.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700157152.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157152.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157152.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157152.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157152.hg.1 |
TC0700008240.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000421579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000421579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000421579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000421579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000441140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000441140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000441140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000454217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000454217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000454217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000454217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157156.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157156.hg.1 |
ENST00000421579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157156.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157156.hg.1 |
ENST00000454217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157156.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157156.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157156.hg.1 |
ENST00000421579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157156.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157156.hg.1 |
ENST00000454217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157156.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157156.hg.1 |
TC0700008240.hg.1 |
Clariom D TC |