Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLIT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271946.1 | NP_001258875.1 | 29 - 30 | 3620 | 104 | |
NM_003062.3 | NP_003053.1 | 29 - 30 | 3599 | 104 | ||
XM_017009779.1 | XP_016865268.1 | 29 - 30 | 3002 | 104 | ||
GenBank mRNA | AB011538.2 | - | 29 - 30 | 3599 | 104 | |
AB017169.1 | - | 29 - 30 | 3442 | 104 | ||
AF075240.1 | - | 13 - 14 | 1460 | 104 | ||
AK001626.2 | - | 5 - 6 | 540 | 104 | ||
AY358884.1 | - | 29 - 30 | 3261 | 104 | ||
BC146759.1 | - | 29 - 30 | 3599 | 9007197 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |