Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABLIM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301015.1 | NP_001287944.1 | 14 - 15 | 1964 | 77 | |
NM_001301018.1 | NP_001287947.1 | 13 - 14 | 1339 | 77 | ||
NM_014945.3 | NP_055760.1 | 15 - 16 | 1610 | 77 | ||
XM_005268392.1 | XP_005268449.1 | 14 - 15 | 1492 | 77 | ||
XM_005268394.1 | XP_005268451.1 | 13 - 14 | 1405 | 77 | ||
XM_005268395.1 | XP_005268452.1 | 12 - 13 | 1306 | 77 | ||
XM_017009238.1 | XP_016864727.1 | 11 - 12 | 1400 | 77 | ||
GenBank mRNA | AB020650.1 | - | 15 - 16 | 1523 | 77 | |
AK225769.1 | - | 14 - 15 | 1832 | 77 | ||
AK289736.1 | - | 14 - 15 | 1377 | 77 | ||
AL833021.1 | - | 8 - 9 | 718 | 77 | ||
DQ413174.1 | - | 13 - 14 | 1301 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons |