Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
ENST00000284262.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
ENST00000284262.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
ENST00000284262.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
ENST00000284262.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
ENST00000284262.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
ENST00000284262.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
ENST00000284262.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
ENST00000284262.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
ENST00000284262.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
ENST00000284262.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
ENST00000301008.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
ENST00000301008.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
ENST00000301008.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
ENST00000537256.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
ENST00000537256.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
ENST00000537256.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
ENST00000537256.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
ENST00000537256.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
ENST00000537256.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
ENST00000537256.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
ENST00000563609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
ENST00000563609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
ENST00000563609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
ENST00000563609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
ENST00000563609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
ENST00000563609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
ENST00000563609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
ENST00000563609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
NM_001271604.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
NM_001271604.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
NM_001271604.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
NM_001271605.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
NM_001271605.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
NM_001271605.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
NM_001271605.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
NM_020655.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
NM_020655.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
NM_020655.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
NM_020655.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
NM_020655.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
NM_020655.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
NM_020655.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
NM_020655.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
NM_020655.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
NM_020655.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
NR_073379.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
NR_073379.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
NR_073379.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
NR_073379.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
NR_073379.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
NR_073379.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
NR_073379.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008779.hg.1 |
NR_073379.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600165451.hg.1 |
ENST00000284262.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600165451.hg.1 |
ENST00000537256.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600165451.hg.1 |
ENST00000563609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600165451.hg.1 |
NM_020655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600165451.hg.1 |
NR_073379.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600165451.hg.1 |
ENST00000284262.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600165451.hg.1 |
ENST00000537256.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600165451.hg.1 |
ENST00000563609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600165451.hg.1 |
NM_020655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600165451.hg.1 |
NR_073379.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600165451.hg.1 |
TC1600008779.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600165452.hg.1 |
ENST00000284262.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600165452.hg.1 |
ENST00000563609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600165452.hg.1 |
NM_020655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600165452.hg.1 |
NR_073379.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600165452.hg.1 |
ENST00000284262.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600165452.hg.1 |
ENST00000563609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600165452.hg.1 |
NM_020655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600165452.hg.1 |
NR_073379.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600165452.hg.1 |
TC1600008779.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
ENST00000284262.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
ENST00000284262.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
ENST00000284262.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
ENST00000284262.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
ENST00000284262.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
ENST00000284262.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
ENST00000284262.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
ENST00000284262.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
ENST00000284262.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
ENST00000284262.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
ENST00000301008.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
ENST00000301008.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
ENST00000301008.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
ENST00000537256.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
ENST00000537256.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
ENST00000537256.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
ENST00000537256.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
ENST00000537256.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
ENST00000537256.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
ENST00000537256.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
ENST00000563609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
ENST00000563609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
ENST00000563609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
ENST00000563609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
ENST00000563609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
ENST00000563609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
ENST00000563609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
ENST00000563609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
NM_001271604.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
NM_001271604.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
NM_001271604.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
NM_001271605.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
NM_001271605.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
NM_001271605.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
NM_001271605.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
NM_020655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
NM_020655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
NM_020655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
NM_020655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
NM_020655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
NM_020655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
NM_020655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
NM_020655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
NM_020655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
NM_020655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
NR_073379.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
NR_073379.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
NR_073379.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
NR_073379.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
NR_073379.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
NR_073379.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
NR_073379.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008779.hg.1 |
NR_073379.2 |
Refseq |