Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZZEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015113.3 | NP_055928.3 | 45 - 46 | 7470 | 92 | |
XM_005256561.2 | XP_005256618.1 | 45 - 46 | 7534 | 92 | ||
XM_017024382.1 | XP_016879871.1 | 45 - 46 | 7534 | 92 | ||
XM_017024383.1 | XP_016879872.1 | 45 - 46 | 7531 | 92 | ||
XM_017024384.1 | XP_016879873.1 | 20 - 21 | 3345 | 92 | ||
GenBank mRNA | AB007859.4 | - | 45 - 46 | 7470 | 92 | |
BC067099.1 | - | 2 - 3 | 228 | 92 | ||
BC151836.1 | - | 45 - 46 | 7470 | 8860131 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |