Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUSC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105204.1 | NP_001098674.1 | 6 - 7 | 2046 | 87 | |
NM_001278229.1 | NP_001265158.1 | 5 - 6 | 1111 | 87 | ||
NR_103478.1 | 4 - 5 | 776 | 87 | |||
XM_006711256.1 | XP_006711319.1 | 7 - 8 | 2457 | 87 | ||
XM_017000892.1 | XP_016856381.1 | 5 - 6 | 725 | 87 | ||
GenBank mRNA | AK074485.1 | - | 4 - 5 | 664 | 87 | |
AK125378.1 | - | 6 - 7 | 1998 | 87 | ||
BC004293.1 | - | 4 - 5 | 632 | 87 | ||
BX402179.2 | - | 5 - 6 | 602 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |