Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000251.hg.1 |
ENST00000246032.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000251.hg.1 |
ENST00000381482.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000251.hg.1 |
ENST00000400064.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000251.hg.1 |
NM_080836.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000251.hg.1 |
TC20000021.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000247.hg.1 |
ENST00000246032.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000247.hg.1 |
ENST00000381482.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000247.hg.1 |
NM_080836.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000247.hg.1 |
TC20000021.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000249.hg.1 |
ENST00000246032.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000249.hg.1 |
ENST00000381482.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000249.hg.1 |
NM_080836.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000249.hg.1 |
TC20000021.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006504.hg.1 |
ENST00000381482.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006504.hg.1 |
ENST00000381482.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006504.hg.1 |
ENST00000381482.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006504.hg.1 |
ENST00000381482.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006504.hg.1 |
ENST00000381482.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006504.hg.1 |
ENST00000381482.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006504.hg.1 |
ENST00000381482.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006504.hg.1 |
ENST00000381482.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006504.hg.1 |
ENST00000493263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006504.hg.1 |
ENST00000493263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006504.hg.1 |
ENST00000493263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006504.hg.1 |
ENST00000493263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006504.hg.1 |
ENST00000493263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006504.hg.1 |
NM_080836.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006504.hg.1 |
NM_080836.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006504.hg.1 |
NM_080836.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006504.hg.1 |
NM_080836.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006504.hg.1 |
NM_080836.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006504.hg.1 |
NM_080836.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006504.hg.1 |
NM_080836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000144554.hg.1 |
ENST00000381482.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000144554.hg.1 |
NM_080836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000144554.hg.1 |
TC2000006504.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000144554.hg.1 |
ENST00000381482.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000144554.hg.1 |
NM_080836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000144557.hg.1 |
ENST00000381482.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000144557.hg.1 |
ENST00000493263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000144557.hg.1 |
NM_080836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000144557.hg.1 |
TC2000006504.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000144557.hg.1 |
ENST00000381482.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000144557.hg.1 |
ENST00000493263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000144557.hg.1 |
NM_080836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000144556.hg.1 |
ENST00000381482.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000144556.hg.1 |
NM_080836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000144556.hg.1 |
TC2000006504.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000144556.hg.1 |
ENST00000381482.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000144556.hg.1 |
NM_080836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006504.hg.1 |
ENST00000381482.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006504.hg.1 |
ENST00000381482.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006504.hg.1 |
ENST00000381482.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006504.hg.1 |
ENST00000381482.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006504.hg.1 |
ENST00000381482.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006504.hg.1 |
ENST00000381482.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006504.hg.1 |
ENST00000381482.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006504.hg.1 |
ENST00000381482.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006504.hg.1 |
ENST00000493263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006504.hg.1 |
ENST00000493263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006504.hg.1 |
ENST00000493263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006504.hg.1 |
ENST00000493263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006504.hg.1 |
ENST00000493263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006504.hg.1 |
NM_080836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006504.hg.1 |
NM_080836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006504.hg.1 |
NM_080836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006504.hg.1 |
NM_080836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006504.hg.1 |
NM_080836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006504.hg.1 |
NM_080836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006504.hg.1 |
NM_080836.3 |
Refseq |