Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSTPIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024430.3 | NP_077748.3 | 6 - 7 | 509 | 77 | |
XM_011526253.2 | XP_011524555.1 | 5 - 6 | 474 | 77 | ||
XM_011526254.1 | XP_011524556.1 | 6 - 7 | 485 | 77 | ||
XM_011526256.1 | XP_011524558.1 | 5 - 6 | 385 | 77 | ||
GenBank mRNA | AK023100.1 | - | 2 - 3 | 77 | 77 | |
BC035395.1 | - | 6 - 7 | 482 | 4524856 | 77 | |
BP352927.1 | - | 6 - 7 | 509 | 77 | ||
KU178455.1 | - | 6 - 7 | 413 | 77 | ||
KU178456.1 | - | 6 - 7 | 413 | 77 | ||
KU178457.1 | - | 5 - 6 | 335 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |