Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12007499.hg.1 |
ENST00000280379.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12007499.hg.1 |
ENST00000393629.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12007499.hg.1 |
ENST00000393630.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12007499.hg.1 |
ENST00000393632.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12007499.hg.1 |
ENST00000546600.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12007499.hg.1 |
ENST00000552738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12007499.hg.1 |
NM_015026.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12007499.hg.1 |
TC12000556.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200158352.hg.1 |
ENST00000393629.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200158352.hg.1 |
ENST00000393630.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200158352.hg.1 |
ENST00000546600.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200158352.hg.1 |
ENST00000546751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200158352.hg.1 |
ENST00000547095.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200158352.hg.1 |
ENST00000551307.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200158352.hg.1 |
ENST00000552738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200158352.hg.1 |
NM_001278469.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200158352.hg.1 |
NM_001278470.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200158352.hg.1 |
NM_001278471.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200158352.hg.1 |
NM_001278472.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200158352.hg.1 |
NM_015026.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200158352.hg.1 |
ENST00000393629.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200158352.hg.1 |
ENST00000393630.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200158352.hg.1 |
ENST00000546600.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200158352.hg.1 |
ENST00000546751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200158352.hg.1 |
ENST00000547095.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200158352.hg.1 |
ENST00000551307.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200158352.hg.1 |
ENST00000552738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200158352.hg.1 |
NM_001278469.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200158352.hg.1 |
NM_001278470.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200158352.hg.1 |
NM_001278471.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200158352.hg.1 |
NM_001278472.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200158352.hg.1 |
NM_015026.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200158352.hg.1 |
TC1200007959.hg.1 |
Clariom D TC |