Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008923.hg.1 |
ENST00000369422.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008923.hg.1 |
ENST00000369422.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008923.hg.1 |
ENST00000369422.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008923.hg.1 |
ENST00000369422.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008923.hg.1 |
ENST00000369422.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008923.hg.1 |
ENST00000369422.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008923.hg.1 |
ENST00000369422.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008923.hg.1 |
ENST00000369422.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008923.hg.1 |
ENST00000369422.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008923.hg.1 |
ENST00000369422.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008923.hg.1 |
ENST00000369422.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008923.hg.1 |
ENST00000369422.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008923.hg.1 |
ENST00000494328.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008923.hg.1 |
ENST00000494328.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008923.hg.1 |
ENST00000494328.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008923.hg.1 |
NM_058222.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008923.hg.1 |
NM_058222.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008923.hg.1 |
NM_058222.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008923.hg.1 |
NM_058222.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008923.hg.1 |
NM_058222.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008923.hg.1 |
NM_058222.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008923.hg.1 |
NM_058222.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008923.hg.1 |
NM_058222.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008923.hg.1 |
NM_058222.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008923.hg.1 |
NM_058222.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008923.hg.1 |
NM_058222.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008923.hg.1 |
NM_058222.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008923.hg.1 |
NM_058222.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008923.hg.1 |
ENST00000369422.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008923.hg.1 |
ENST00000369422.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008923.hg.1 |
ENST00000369422.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008923.hg.1 |
ENST00000369422.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008923.hg.1 |
ENST00000369422.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008923.hg.1 |
ENST00000369422.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008923.hg.1 |
ENST00000369422.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008923.hg.1 |
ENST00000369422.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008923.hg.1 |
ENST00000369422.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008923.hg.1 |
ENST00000369422.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008923.hg.1 |
ENST00000369422.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008923.hg.1 |
ENST00000369422.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008923.hg.1 |
ENST00000494328.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008923.hg.1 |
ENST00000494328.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008923.hg.1 |
ENST00000494328.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008923.hg.1 |
NM_058222.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008923.hg.1 |
NM_058222.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008923.hg.1 |
NM_058222.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008923.hg.1 |
NM_058222.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008923.hg.1 |
NM_058222.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008923.hg.1 |
NM_058222.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008923.hg.1 |
NM_058222.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008923.hg.1 |
NM_058222.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008923.hg.1 |
NM_058222.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008923.hg.1 |
NM_058222.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008923.hg.1 |
NM_058222.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008923.hg.1 |
NM_058222.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008923.hg.1 |
NM_058222.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161017.hg.1 |
ENST00000369422.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161017.hg.1 |
NM_058222.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161017.hg.1 |
ENST00000369422.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161017.hg.1 |
NM_058222.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161017.hg.1 |
TC1000008923.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000161019.hg.1 |
ENST00000369422.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161019.hg.1 |
NM_058222.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161019.hg.1 |
ENST00000369422.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161019.hg.1 |
NM_058222.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161019.hg.1 |
TC1000008923.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000161016.hg.1 |
ENST00000369422.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161016.hg.1 |
NM_058222.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161016.hg.1 |
ENST00000369422.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161016.hg.1 |
NM_058222.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161016.hg.1 |
TC1000008923.hg.1 |
Clariom D TC |