Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012172.hg.1 |
ENST00000233139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012172.hg.1 |
ENST00000233139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012172.hg.1 |
ENST00000233139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012172.hg.1 |
ENST00000233139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012172.hg.1 |
ENST00000233139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012172.hg.1 |
ENST00000233139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012172.hg.1 |
ENST00000233139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012172.hg.1 |
ENST00000405650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012172.hg.1 |
ENST00000405650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012172.hg.1 |
ENST00000405650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012172.hg.1 |
ENST00000405650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012172.hg.1 |
ENST00000405650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012172.hg.1 |
ENST00000405650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012172.hg.1 |
ENST00000405650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012172.hg.1 |
ENST00000405650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012172.hg.1 |
ENST00000405650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012172.hg.1 |
ENST00000405650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012172.hg.1 |
ENST00000405650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012172.hg.1 |
ENST00000622030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012172.hg.1 |
ENST00000622030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012172.hg.1 |
ENST00000622030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012172.hg.1 |
ENST00000622030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012172.hg.1 |
ENST00000622030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012172.hg.1 |
ENST00000622030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012172.hg.1 |
ENST00000622030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012172.hg.1 |
ENST00000622030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012172.hg.1 |
ENST00000622030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012172.hg.1 |
ENST00000622030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012172.hg.1 |
ENST00000622030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012172.hg.1 |
NM_000348.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012172.hg.1 |
NM_000348.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012172.hg.1 |
NM_000348.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012172.hg.1 |
NM_000348.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012172.hg.1 |
NM_000348.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012172.hg.1 |
NM_000348.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012172.hg.1 |
NM_000348.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012172.hg.1 |
NM_000348.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012172.hg.1 |
NM_000348.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008870.hg.1 |
ENST00000419384.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027703.hg.1 |
TC02001716.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027703.hg.1 |
ENST00000233139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027703.hg.1 |
ENST00000405650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027703.hg.1 |
NM_000348.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200188860.hg.1 |
ENST00000233139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200188860.hg.1 |
ENST00000405650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200188860.hg.1 |
ENST00000622030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200188860.hg.1 |
NM_000348.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200188860.hg.1 |
ENST00000233139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200188860.hg.1 |
ENST00000405650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200188860.hg.1 |
ENST00000622030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200188860.hg.1 |
NM_000348.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200188860.hg.1 |
TC0200012172.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200012172.hg.1 |
ENST00000233139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012172.hg.1 |
ENST00000233139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012172.hg.1 |
ENST00000233139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012172.hg.1 |
ENST00000233139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012172.hg.1 |
ENST00000233139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012172.hg.1 |
ENST00000233139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012172.hg.1 |
ENST00000233139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012172.hg.1 |
ENST00000405650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012172.hg.1 |
ENST00000405650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012172.hg.1 |
ENST00000405650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012172.hg.1 |
ENST00000405650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012172.hg.1 |
ENST00000405650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012172.hg.1 |
ENST00000405650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012172.hg.1 |
ENST00000405650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012172.hg.1 |
ENST00000405650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012172.hg.1 |
ENST00000405650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012172.hg.1 |
ENST00000405650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012172.hg.1 |
ENST00000405650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012172.hg.1 |
ENST00000622030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012172.hg.1 |
ENST00000622030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012172.hg.1 |
ENST00000622030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012172.hg.1 |
ENST00000622030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012172.hg.1 |
ENST00000622030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012172.hg.1 |
ENST00000622030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012172.hg.1 |
ENST00000622030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012172.hg.1 |
ENST00000622030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012172.hg.1 |
ENST00000622030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012172.hg.1 |
ENST00000622030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012172.hg.1 |
ENST00000622030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012172.hg.1 |
NM_000348.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012172.hg.1 |
NM_000348.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012172.hg.1 |
NM_000348.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012172.hg.1 |
NM_000348.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012172.hg.1 |
NM_000348.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012172.hg.1 |
NM_000348.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012172.hg.1 |
NM_000348.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012172.hg.1 |
NM_000348.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012172.hg.1 |
NM_000348.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008870.hg.1 |
ENST00000419384.1 |
Ensembl |