Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012877.hg.1 |
TC06001091.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012877.hg.1 |
ENST00000361131.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012877.hg.1 |
NM_030949.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009831.hg.1 |
ENST00000361131.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009831.hg.1 |
ENST00000361131.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009831.hg.1 |
ENST00000361131.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009831.hg.1 |
ENST00000361131.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009831.hg.1 |
ENST00000361131.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009831.hg.1 |
ENST00000361131.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009831.hg.1 |
ENST00000361131.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009831.hg.1 |
NM_030949.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009831.hg.1 |
NM_030949.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009831.hg.1 |
NM_030949.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009831.hg.1 |
NM_030949.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009831.hg.1 |
NM_030949.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009831.hg.1 |
NM_030949.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009831.hg.1 |
NM_030949.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009831.hg.1 |
NM_030949.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600167276.hg.1 |
ENST00000361131.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167276.hg.1 |
NM_030949.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600167276.hg.1 |
ENST00000361131.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600167276.hg.1 |
NM_030949.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600167276.hg.1 |
TC0600009831.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600009831.hg.1 |
ENST00000361131.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009831.hg.1 |
ENST00000361131.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009831.hg.1 |
ENST00000361131.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009831.hg.1 |
ENST00000361131.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009831.hg.1 |
ENST00000361131.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009831.hg.1 |
ENST00000361131.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009831.hg.1 |
ENST00000361131.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009831.hg.1 |
NM_030949.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009831.hg.1 |
NM_030949.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009831.hg.1 |
NM_030949.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009831.hg.1 |
NM_030949.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009831.hg.1 |
NM_030949.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009831.hg.1 |
NM_030949.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009831.hg.1 |
NM_030949.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009831.hg.1 |
NM_030949.2 |
Refseq |