Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035730.hg.1 |
TC01002326.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035730.hg.1 |
ENST00000290101.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035730.hg.1 |
ENST00000374758.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035730.hg.1 |
ENST00000374761.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035730.hg.1 |
ENST00000374763.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035730.hg.1 |
ENST00000374765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035730.hg.1 |
ENST00000487268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035730.hg.1 |
ENST00000542643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035730.hg.1 |
NM_001145657.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035730.hg.1 |
NM_001145658.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035730.hg.1 |
NM_002885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197943.hg.1 |
ENST00000290101.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197943.hg.1 |
ENST00000374761.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197943.hg.1 |
ENST00000374763.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197943.hg.1 |
ENST00000374765.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197943.hg.1 |
ENST00000471600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197943.hg.1 |
ENST00000495204.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197943.hg.1 |
ENST00000542643.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197943.hg.1 |
ENST00000611549.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197943.hg.1 |
NM_001145657.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197943.hg.1 |
NM_001145658.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197943.hg.1 |
NM_002885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197943.hg.1 |
ENST00000290101.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197943.hg.1 |
ENST00000374761.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197943.hg.1 |
ENST00000374763.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197943.hg.1 |
ENST00000374765.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197943.hg.1 |
ENST00000471600.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197943.hg.1 |
ENST00000495204.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197943.hg.1 |
ENST00000542643.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197943.hg.1 |
ENST00000611549.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197943.hg.1 |
NM_001145657.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197943.hg.1 |
NM_001145658.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197943.hg.1 |
NM_002885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197943.hg.1 |
TC0100013223.hg.1 |
Clariom D TC |