Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001184997.1 | NP_001171926.1 | 1 - 2 | 103 | 105 | |
XM_011515804.2 | XP_011514106.1 | 2 - 3 | 167 | 105 | ||
XM_011515805.2 | XP_011514107.1 | 2 - 3 | 324 | 105 | ||
GenBank mRNA | AA412402.1 | - | 1 - 2 | 113 | 105 | |
AF325690.1 | - | 1 - 2 | 53 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |