Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007161.3 | NP_009092.3 | 2 - 3 | 216 | 111 | |
NM_205839.2 | NP_995311.2 | 3 - 4 | 353 | 90 | ||
XM_006715206.3 | XP_006715269.1 | 3 - 4 | 345 | 111 | ||
XM_011514914.2 | XP_011513216.1 | 3 - 4 | 326 | 111 | ||
GenBank mRNA | AF000425.1 | - | 3 - 4 | 331 | 90 | |
AY852266.1 | - | 2 - 3 | 116 | 111 | ||
CD742958.1 | - | 2 - 3 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Inventoried Probe spans exons |