Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000272937.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000272937.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000272937.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000272937.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000272937.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000272937.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000272937.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000272937.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000272937.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000272937.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000272937.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000272937.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000272937.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000272937.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000272937.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409002.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409002.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409002.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409002.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409002.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409002.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409002.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409002.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409002.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409002.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409002.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409002.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409002.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409160.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409160.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409160.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409160.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409160.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409160.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409160.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409160.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409160.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409160.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409160.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409160.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409160.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409160.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409160.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409574.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409574.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409574.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409574.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409574.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409574.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409574.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409574.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000409574.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000417803.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000417803.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000417803.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000417803.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000417803.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000417803.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000436051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000436051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000436051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000436051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000436051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000436051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000450098.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000450098.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000450098.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000450098.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000450098.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000450098.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
ENST00000450098.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_001142853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_001142853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_001142853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_001142853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_001142853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_001142853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_001142853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_001142853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_001142853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_001142853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_001142853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_001142853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_001142853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_001142853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_001142853.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_001282434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_001282434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_001282434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_001282434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_001282434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_001282434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_001282434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_001282434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_001282434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_001282434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_001282434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_001282434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_001282434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_001282434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_018645.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_018645.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_018645.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_018645.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_018645.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_018645.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_018645.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_018645.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_018645.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_018645.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_018645.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_018645.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_018645.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_018645.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016222.hg.1 |
NM_018645.5 |
Refseq |