Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000257604.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000257604.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000257604.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000257604.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000257604.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000257604.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000257604.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000257604.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000257604.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000257604.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000257604.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000257604.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000257604.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000257604.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000257604.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000257604.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000257604.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000257604.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000257604.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000257604.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000412615.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000412615.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000412615.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000412615.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000412615.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000412615.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000412615.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000412615.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000412615.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000412615.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000412615.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000412615.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000412615.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000412615.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000412615.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000412615.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000412615.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000412615.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000412615.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000547063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000547063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000547063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000547063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000547063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000547063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000547063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000547063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000547063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000548092.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000548092.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000548092.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000548092.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000548092.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000548092.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000548277.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000548277.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000548277.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000548277.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000548277.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000549358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000549358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000549358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000549358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000549358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000549358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000549358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000550051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000550051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000550051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000550051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000550051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000550051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000550051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000550051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000552890.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000552890.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000552890.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000552890.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000552890.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000552890.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000552890.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000552890.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000552890.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000552890.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000552890.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000552890.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000552896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000552896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000552896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000552896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000552896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000552896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000552896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000552896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000552896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
ENST00000552896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
NM_001143906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
NM_001143906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
NM_001143906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
NM_001143906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
NM_001143906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
NM_001143906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
NM_001143906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
NM_001143906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
NM_001143906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
NM_001143906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
NM_001143906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
NM_001143906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
NM_001143906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
NM_001143906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
NM_001143906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
NM_001143906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
NM_001143906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
NM_001143906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
NM_001143906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
NM_001143906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
NM_006700.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
NM_006700.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
NM_006700.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
NM_006700.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
NM_006700.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
NM_006700.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
NM_006700.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
NM_006700.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
NM_006700.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
NM_006700.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
NM_006700.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
NM_006700.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
NM_006700.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
NM_006700.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
NM_006700.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
NM_006700.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
NM_006700.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008906.hg.1 |
NM_006700.2 |
Refseq |