Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001184993.1 | NP_001171922.1 | 30 - 31 | 4190 | 256 | |
NM_031277.2 | NP_112567.2 | 30 - 31 | 4202 | 256 | ||
XM_006719846.3 | XP_006719909.1 | 31 - 32 | 4643 | 256 | ||
XM_006719849.2 | XP_006719912.1 | 31 - 32 | 4360 | 256 | ||
XM_006719852.2 | XP_006719915.1 | 30 - 31 | 4238 | 256 | ||
XM_011535152.2 | XP_011533454.1 | 35 - 36 | 6268 | 256 | ||
XM_011535155.2 | XP_011533457.1 | 34 - 35 | 5004 | 256 | ||
XM_011535156.2 | XP_011533458.1 | 35 - 36 | 6268 | 256 | ||
XM_011535157.2 | XP_011533459.1 | 35 - 36 | 6256 | 256 | ||
XM_011535158.2 | XP_011533460.1 | 34 - 35 | 6142 | 130 | ||
XM_011535159.2 | XP_011533461.1 | 34 - 35 | 6121 | 256 | ||
XM_011535160.2 | XP_011533462.1 | 34 - 35 | 6109 | 256 | ||
XM_011535162.1 | XP_011533464.1 | 27 - 28 | 3884 | 256 | ||
XM_011535163.1 | XP_011533465.1 | 22 - 23 | 3154 | 256 | ||
XM_011535164.2 | XP_011533466.1 | 16 - 17 | 2050 | 256 | ||
XM_017020676.1 | XP_016876165.1 | 31 - 32 | 4312 | 256 | ||
GenBank mRNA | AK001907.1 | - | 10 - 11 | 1297 | 130 | |
BC146822.1 | - | 31 - 32 | 4180 | 256 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |