Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006653.hg.1 |
ENST00000225973.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006653.hg.1 |
ENST00000544533.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006653.hg.1 |
NM_005440.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006653.hg.1 |
TC17000546.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007934.hg.1 |
ENST00000225973.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007934.hg.1 |
ENST00000587117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007934.hg.1 |
ENST00000587117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007934.hg.1 |
ENST00000587117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007934.hg.1 |
ENST00000587117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007934.hg.1 |
ENST00000587250.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007934.hg.1 |
ENST00000587250.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007934.hg.1 |
ENST00000587250.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007934.hg.1 |
ENST00000587250.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007934.hg.1 |
ENST00000587250.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007934.hg.1 |
ENST00000587250.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007934.hg.1 |
ENST00000587250.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007934.hg.1 |
ENST00000587250.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007934.hg.1 |
ENST00000587250.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007934.hg.1 |
NM_005440.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007934.hg.1 |
NM_005440.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007934.hg.1 |
NM_005440.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007934.hg.1 |
NM_005440.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007934.hg.1 |
NM_005440.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007934.hg.1 |
NM_005440.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007934.hg.1 |
NM_005440.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007934.hg.1 |
NM_005440.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007934.hg.1 |
NM_005440.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006655.hg.1 |
ENST00000225973.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006655.hg.1 |
ENST00000544533.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006655.hg.1 |
NM_005440.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006655.hg.1 |
TC17000546.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006654.hg.1 |
ENST00000225973.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006654.hg.1 |
ENST00000544533.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006654.hg.1 |
NM_005440.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006654.hg.1 |
TC17000546.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700157993.hg.1 |
ENST00000587250.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157993.hg.1 |
NM_005440.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700157993.hg.1 |
ENST00000587250.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157993.hg.1 |
NM_005440.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700157993.hg.1 |
TC1700007934.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700157997.hg.1 |
ENST00000225973.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157997.hg.1 |
ENST00000587250.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157997.hg.1 |
NM_005440.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700157997.hg.1 |
ENST00000225973.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157997.hg.1 |
ENST00000587250.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157997.hg.1 |
NM_005440.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700157997.hg.1 |
TC1700007934.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700157995.hg.1 |
ENST00000587250.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157995.hg.1 |
NM_005440.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700157995.hg.1 |
ENST00000587250.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157995.hg.1 |
NM_005440.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700157995.hg.1 |
TC1700007934.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700157996.hg.1 |
ENST00000587250.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157996.hg.1 |
NM_005440.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700157996.hg.1 |
ENST00000587250.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157996.hg.1 |
NM_005440.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700157996.hg.1 |
TC1700007934.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700007934.hg.1 |
ENST00000225973.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007934.hg.1 |
ENST00000587117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007934.hg.1 |
ENST00000587117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007934.hg.1 |
ENST00000587117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007934.hg.1 |
ENST00000587117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007934.hg.1 |
ENST00000587250.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007934.hg.1 |
ENST00000587250.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007934.hg.1 |
ENST00000587250.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007934.hg.1 |
ENST00000587250.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007934.hg.1 |
ENST00000587250.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007934.hg.1 |
ENST00000587250.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007934.hg.1 |
ENST00000587250.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007934.hg.1 |
ENST00000587250.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007934.hg.1 |
ENST00000587250.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007934.hg.1 |
NM_005440.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007934.hg.1 |
NM_005440.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007934.hg.1 |
NM_005440.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007934.hg.1 |
NM_005440.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007934.hg.1 |
NM_005440.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007934.hg.1 |
NM_005440.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007934.hg.1 |
NM_005440.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007934.hg.1 |
NM_005440.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007934.hg.1 |
NM_005440.4 |
Refseq |