Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRPPRC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133259.3 | NP_573566.2 | 35 - 36 | 3958 | 129 | |
XM_006711915.2 | XP_006711978.1 | 35 - 36 | 3967 | 129 | ||
GenBank mRNA | AK125781.1 | - | 35 - 36 | 3929 | 129 | |
AK130686.1 | - | 4 - 5 | 2943 | 129 | ||
AK290016.1 | - | 35 - 36 | 3929 | 129 | ||
AY289212.1 | - | 35 - 36 | 3958 | 129 | ||
BC010282.1 | - | 15 - 16 | 1825 | 129 | ||
BC050311.1 | - | 35 - 36 | 3909 | 4827456 | 129 | |
BC130285.1 | - | 35 - 36 | 3954 | 40146316 | 129 | |
M92439.1 | - | 34 - 35 | 3582 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |