Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003248.hg.1 |
ENST00000305560.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003248.hg.1 |
ENST00000559860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003248.hg.1 |
NM_024063.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003248.hg.1 |
NR_027635.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003248.hg.1 |
TC15000356.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000305560.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000305560.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000305560.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000305560.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000305560.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000305560.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000305560.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000305560.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000305560.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000305560.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000305560.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000305560.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000305560.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000305560.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000305560.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000305560.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000525552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000525552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000525552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000531624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000531624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000531624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000531624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000531624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000531624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000531624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000531624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000531624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000531970.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000531970.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000531970.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000531970.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000531970.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000531970.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000531970.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000531970.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000531970.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000531970.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000531970.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000531970.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000531970.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000531970.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000533199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000533199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000533199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000533841.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000533841.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000533841.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000533841.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000533841.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000559860.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000559860.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000559860.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000559860.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000559860.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000559860.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000559860.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000559860.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000559860.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
ENST00000559860.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
NM_024063.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
NM_024063.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
NM_024063.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
NM_024063.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
NM_024063.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
NM_024063.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
NM_024063.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
NM_024063.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
NM_024063.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
NM_024063.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
NM_024063.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
NM_024063.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
NM_024063.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
NM_024063.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
NM_024063.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
NM_024063.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
NR_027635.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
NR_027635.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
NR_027635.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
NR_027635.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
NR_027635.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
NR_027635.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
NR_027635.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
NR_027635.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
NR_027635.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
NR_027635.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
NR_027635.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
NR_027635.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
NR_027635.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
NR_027635.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
NR_027635.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
NR_027635.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007121.hg.1 |
NR_027635.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000305560.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000305560.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000305560.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000305560.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000305560.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000305560.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000305560.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000305560.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000305560.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000305560.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000305560.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000305560.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000305560.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000305560.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000305560.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000305560.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000525552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000525552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000525552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000531624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000531624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000531624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000531624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000531624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000531624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000531624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000531624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000531624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000531970.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000531970.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000531970.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000531970.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000531970.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000531970.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000531970.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000531970.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000531970.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000531970.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000531970.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000531970.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000531970.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000531970.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000533199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000533199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000533199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000533841.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000533841.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000533841.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000533841.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000533841.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000559860.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000559860.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000559860.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000559860.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000559860.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000559860.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000559860.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000559860.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000559860.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
ENST00000559860.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
NM_024063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
NM_024063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
NM_024063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
NM_024063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
NM_024063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
NM_024063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
NM_024063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
NM_024063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
NM_024063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
NM_024063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
NM_024063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
NM_024063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
NM_024063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
NM_024063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
NM_024063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
NM_024063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
NR_027635.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
NR_027635.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
NR_027635.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
NR_027635.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
NR_027635.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
NR_027635.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
NR_027635.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
NR_027635.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
NR_027635.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
NR_027635.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
NR_027635.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
NR_027635.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
NR_027635.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
NR_027635.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
NR_027635.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
NR_027635.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007121.hg.1 |
NR_027635.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500150660.hg.1 |
ENST00000305560.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500150660.hg.1 |
ENST00000531624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500150660.hg.1 |
ENST00000531970.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500150660.hg.1 |
ENST00000559860.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500150660.hg.1 |
NM_024063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500150660.hg.1 |
NR_027635.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500150660.hg.1 |
ENST00000305560.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500150660.hg.1 |
ENST00000531624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500150660.hg.1 |
ENST00000531970.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500150660.hg.1 |
ENST00000559860.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500150660.hg.1 |
NM_024063.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500150660.hg.1 |
NR_027635.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500150660.hg.1 |
TC1500007121.hg.1 |
Clariom D TC |