Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034915.2 | NP_001030087.2 | 15 - 16 | 2243 | 71 | |
NM_017697.3 | NP_060167.2 | 15 - 16 | 2255 | 71 | ||
XM_005250992.3 | XP_005251049.1 | 14 - 15 | 2241 | 71 | ||
GenBank mRNA | BC035406.1 | - | 11 - 12 | 1517 | 71 | |
BC067098.1 | - | 15 - 16 | 2209 | 6586195 | 71 | |
BC099916.1 | - | 15 - 16 | 2207 | 30527936 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons |