Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020135.hg.1 |
TC08001532.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020135.hg.1 |
ENST00000297404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020135.hg.1 |
ENST00000524391.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020135.hg.1 |
ENST00000545728.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020135.hg.1 |
NM_014379.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008554.hg.1 |
ENST00000530667.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008554.hg.1 |
ENST00000530667.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011486.hg.1 |
ENST00000297404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011486.hg.1 |
ENST00000297404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011486.hg.1 |
ENST00000297404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011486.hg.1 |
ENST00000297404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011486.hg.1 |
ENST00000297404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011486.hg.1 |
ENST00000297404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011486.hg.1 |
ENST00000297404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011486.hg.1 |
ENST00000524391.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011486.hg.1 |
ENST00000524391.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011486.hg.1 |
ENST00000524391.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011486.hg.1 |
ENST00000524391.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011486.hg.1 |
ENST00000524391.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011486.hg.1 |
ENST00000524391.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011486.hg.1 |
ENST00000524391.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011486.hg.1 |
ENST00000524391.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011486.hg.1 |
ENST00000524391.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011486.hg.1 |
ENST00000524391.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011486.hg.1 |
ENST00000524391.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011486.hg.1 |
NM_014379.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011486.hg.1 |
NM_014379.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011486.hg.1 |
NM_014379.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011486.hg.1 |
NM_014379.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011486.hg.1 |
NM_014379.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011486.hg.1 |
NM_014379.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011486.hg.1 |
NM_014379.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800178183.hg.1 |
ENST00000297404.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178183.hg.1 |
ENST00000524391.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178183.hg.1 |
NM_014379.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800178183.hg.1 |
ENST00000297404.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178183.hg.1 |
ENST00000524391.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178183.hg.1 |
NM_014379.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800178183.hg.1 |
TC0800011486.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800011486.hg.1 |
ENST00000297404.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011486.hg.1 |
ENST00000297404.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011486.hg.1 |
ENST00000297404.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011486.hg.1 |
ENST00000297404.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011486.hg.1 |
ENST00000297404.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011486.hg.1 |
ENST00000297404.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011486.hg.1 |
ENST00000297404.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011486.hg.1 |
ENST00000524391.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011486.hg.1 |
ENST00000524391.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011486.hg.1 |
ENST00000524391.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011486.hg.1 |
ENST00000524391.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011486.hg.1 |
ENST00000524391.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011486.hg.1 |
ENST00000524391.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011486.hg.1 |
ENST00000524391.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011486.hg.1 |
ENST00000524391.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011486.hg.1 |
ENST00000524391.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011486.hg.1 |
ENST00000524391.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011486.hg.1 |
ENST00000524391.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011486.hg.1 |
NM_014379.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011486.hg.1 |
NM_014379.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011486.hg.1 |
NM_014379.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011486.hg.1 |
NM_014379.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011486.hg.1 |
NM_014379.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011486.hg.1 |
NM_014379.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011486.hg.1 |
NM_014379.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008554.hg.1 |
ENST00000530667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008554.hg.1 |
ENST00000530667.1 |
Ensembl |