Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017020612.1 | XP_016876101.1 | 23 | 4394 | 145 | |
GenBank mRNA | AL122056.1 | - | 1 | 3040 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |