Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005572.hg.1 |
ENST00000534842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005572.hg.1 |
ENST00000546655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005572.hg.1 |
ENST00000551660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005572.hg.1 |
NM_001170790.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005572.hg.1 |
TC12000433.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005576.hg.1 |
ENST00000328704.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005576.hg.1 |
ENST00000534842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005576.hg.1 |
ENST00000551660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005576.hg.1 |
NM_001170790.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005576.hg.1 |
TC12000433.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
ENST00000329548.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
ENST00000329548.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
ENST00000329548.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
ENST00000329548.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
ENST00000329548.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
ENST00000329548.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
ENST00000534842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
ENST00000534842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
ENST00000534842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
ENST00000534842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
ENST00000534842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
ENST00000534842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
ENST00000534842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
ENST00000534842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
ENST00000534842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
ENST00000534842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
ENST00000546655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
ENST00000546655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
ENST00000546655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
ENST00000546655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
ENST00000551660.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
ENST00000551660.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
ENST00000551660.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
ENST00000551660.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
ENST00000551660.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
ENST00000551660.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
ENST00000552097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
ENST00000552097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
NM_001170790.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
NM_001170790.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
NM_001170790.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
NM_001170790.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
NM_001170790.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
NM_001170790.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
NM_001170790.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
NM_001170790.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
NM_001170790.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
NM_001170790.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
NM_032889.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
NM_032889.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
NM_032889.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
NM_032889.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
NM_032889.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007706.hg.1 |
NM_032889.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005573.hg.1 |
ENST00000534842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005573.hg.1 |
ENST00000551660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005573.hg.1 |
NM_001170790.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005573.hg.1 |
TC12000433.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005574.hg.1 |
ENST00000328704.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005574.hg.1 |
ENST00000534842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005574.hg.1 |
ENST00000551660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005574.hg.1 |
NM_001170790.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005574.hg.1 |
TC12000433.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005575.hg.1 |
ENST00000328704.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005575.hg.1 |
ENST00000534842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005575.hg.1 |
ENST00000546655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005575.hg.1 |
ENST00000551660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005575.hg.1 |
NM_001170790.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005575.hg.1 |
TC12000433.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200155065.hg.1 |
ENST00000534842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155065.hg.1 |
ENST00000551660.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155065.hg.1 |
NM_001170790.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200155065.hg.1 |
ENST00000534842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155065.hg.1 |
ENST00000551660.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155065.hg.1 |
NM_001170790.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200155065.hg.1 |
TC1200007706.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200155066.hg.1 |
ENST00000534842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155066.hg.1 |
ENST00000546655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155066.hg.1 |
ENST00000551660.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155066.hg.1 |
NM_001170790.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200155066.hg.1 |
ENST00000534842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155066.hg.1 |
ENST00000546655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155066.hg.1 |
ENST00000551660.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155066.hg.1 |
NM_001170790.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200155066.hg.1 |
TC1200007706.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200155064.hg.1 |
ENST00000534842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155064.hg.1 |
NM_001170790.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200155064.hg.1 |
ENST00000534842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155064.hg.1 |
NM_001170790.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200155064.hg.1 |
TC1200007706.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
ENST00000329548.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
ENST00000329548.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
ENST00000329548.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
ENST00000329548.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
ENST00000329548.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
ENST00000329548.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
ENST00000534842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
ENST00000534842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
ENST00000534842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
ENST00000534842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
ENST00000534842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
ENST00000534842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
ENST00000534842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
ENST00000534842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
ENST00000534842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
ENST00000534842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
ENST00000546655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
ENST00000546655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
ENST00000546655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
ENST00000546655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
ENST00000551660.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
ENST00000551660.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
ENST00000551660.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
ENST00000551660.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
ENST00000551660.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
ENST00000551660.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
ENST00000552097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
ENST00000552097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
NM_001170790.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
NM_001170790.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
NM_001170790.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
NM_001170790.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
NM_001170790.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
NM_001170790.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
NM_001170790.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
NM_001170790.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
NM_001170790.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
NM_001170790.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
NM_032889.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
NM_032889.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
NM_032889.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
NM_032889.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
NM_032889.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007706.hg.1 |
NM_032889.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200155063.hg.1 |
ENST00000534842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155063.hg.1 |
ENST00000546655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155063.hg.1 |
NM_001170790.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200155063.hg.1 |
ENST00000534842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155063.hg.1 |
ENST00000546655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155063.hg.1 |
NM_001170790.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200155063.hg.1 |
TC1200007706.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200155067.hg.1 |
ENST00000534842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155067.hg.1 |
ENST00000546655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155067.hg.1 |
ENST00000551660.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155067.hg.1 |
NM_001170790.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200155067.hg.1 |
ENST00000534842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155067.hg.1 |
ENST00000546655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155067.hg.1 |
ENST00000551660.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155067.hg.1 |
NM_001170790.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200155067.hg.1 |
TC1200007706.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200155068.hg.1 |
ENST00000534842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155068.hg.1 |
ENST00000551660.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155068.hg.1 |
NM_001170790.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200155068.hg.1 |
ENST00000534842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155068.hg.1 |
ENST00000551660.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155068.hg.1 |
NM_001170790.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200155068.hg.1 |
TC1200007706.hg.1 |
Clariom D TC |