Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005666.hg.1 |
ENST00000394009.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005666.hg.1 |
ENST00000489323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005666.hg.1 |
NM_025057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005666.hg.1 |
TC14000454.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000394009.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000394009.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000394009.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000394009.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000394009.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000394009.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000394009.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000394009.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000394009.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000394009.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000394009.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000394009.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000394009.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000394009.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000394009.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000463558.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000463558.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000463558.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000463558.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000463558.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000463558.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000464394.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000464394.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000464394.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000464394.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000464394.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000464394.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000464394.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000464394.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000464394.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000477986.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000477986.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000477986.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000477986.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000477986.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000489323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000489323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000489323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000489323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000489323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000489323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000489323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000489323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000492247.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000492247.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000492247.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000492247.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
ENST00000492247.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
NM_025057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
NM_025057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
NM_025057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
NM_025057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
NM_025057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
NM_025057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
NM_025057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
NM_025057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
NM_025057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
NM_025057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
NM_025057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
NM_025057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
NM_025057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
NM_025057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007665.hg.1 |
NM_025057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005667.hg.1 |
ENST00000394009.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005667.hg.1 |
ENST00000489323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005667.hg.1 |
NM_025057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005667.hg.1 |
TC14000454.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1400154090.hg.1 |
ENST00000394009.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154090.hg.1 |
ENST00000464394.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154090.hg.1 |
ENST00000477986.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154090.hg.1 |
ENST00000489323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154090.hg.1 |
NM_025057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154090.hg.1 |
ENST00000394009.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154090.hg.1 |
ENST00000464394.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154090.hg.1 |
ENST00000477986.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154090.hg.1 |
ENST00000489323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154090.hg.1 |
NM_025057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154090.hg.1 |
TC1400007665.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400154089.hg.1 |
ENST00000394009.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154089.hg.1 |
ENST00000464394.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154089.hg.1 |
ENST00000489323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154089.hg.1 |
NM_025057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154089.hg.1 |
ENST00000394009.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154089.hg.1 |
ENST00000464394.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154089.hg.1 |
ENST00000489323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154089.hg.1 |
NM_025057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154089.hg.1 |
TC1400007665.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000394009.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000394009.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000394009.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000394009.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000394009.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000394009.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000394009.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000394009.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000394009.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000394009.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000394009.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000394009.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000394009.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000394009.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000394009.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000463558.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000463558.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000463558.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000463558.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000463558.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000463558.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000464394.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000464394.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000464394.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000464394.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000464394.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000464394.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000464394.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000464394.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000464394.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000477986.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000477986.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000477986.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000477986.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000477986.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000489323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000489323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000489323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000489323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000489323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000489323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000489323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000489323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000492026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000492247.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000492247.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000492247.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000492247.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
ENST00000492247.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
NM_025057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
NM_025057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
NM_025057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
NM_025057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
NM_025057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
NM_025057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
NM_025057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
NM_025057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
NM_025057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
NM_025057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
NM_025057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
NM_025057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
NM_025057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
NM_025057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007665.hg.1 |
NM_025057.2 |
Refseq |