Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNC2H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080463.1 | NP_001073932.1 | 71 - 72 | 10857 | 146 | |
NM_001377.2 | NP_001368.2 | 70 - 71 | 10836 | 146 | ||
XM_006718903.2 | XP_006718966.1 | 70 - 71 | 10815 | 146 | ||
XM_017018291.1 | XP_016873780.1 | 70 - 71 | 10836 | 146 | ||
XM_017018292.1 | XP_016873781.1 | 67 - 68 | 10090 | 146 | ||
GenBank mRNA | AB082528.2 | - | 22 - 23 | 2827 | 146 | |
AB290167.1 | - | 70 - 71 | 10890 | 146 | ||
AK095579.1 | - | 5 - 6 | 563 | 146 | ||
AK131453.1 | - | 15 - 16 | 1830 | 146 | ||
BC037496.1 | - | 7 - 8 | 828 | 146 | ||
BX538093.1 | - | 24 - 25 | 2967 | 146 | ||
BX649161.1 | - | 18 - 19 | 2211 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |