Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNC2H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080463.1 | NP_001073932.1 | 79 - 80 | 11659 | 138 | |
NM_001377.2 | NP_001368.2 | 78 - 79 | 11638 | 138 | ||
XM_006718903.2 | XP_006718966.1 | 78 - 79 | 11617 | 138 | ||
XM_017018291.1 | XP_016873780.1 | 78 - 79 | 11638 | 138 | ||
XM_017018292.1 | XP_016873781.1 | 75 - 76 | 10892 | 138 | ||
GenBank mRNA | AB082528.2 | - | 30 - 31 | 3629 | 138 | |
AB290167.1 | - | 78 - 79 | 11692 | 138 | ||
AK021818.1 | - | 3 - 4 | 270 | 138 | ||
AK095579.1 | - | 13 - 14 | 1365 | 138 | ||
AK131453.1 | - | 23 - 24 | 2632 | 138 | ||
BC037496.1 | - | 15 - 16 | 1630 | 138 | ||
BC038344.1 | - | 5 - 6 | 569 | 138 | ||
BX538093.1 | - | 32 - 33 | 3769 | 138 | ||
BX649161.1 | - | 26 - 27 | 3013 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |