Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027736.hg.1 |
ENST00000260210.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027736.hg.1 |
ENST00000375445.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027736.hg.1 |
NM_001159736.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027736.hg.1 |
NM_032725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027736.hg.1 |
TC11002322.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
ENST00000260210.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
ENST00000260210.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
ENST00000260210.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
ENST00000260210.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
ENST00000260210.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
ENST00000260210.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
ENST00000260210.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
ENST00000260210.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
ENST00000260210.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
ENST00000260210.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
ENST00000260210.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
ENST00000260210.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
ENST00000260210.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
ENST00000375445.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
ENST00000375445.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
ENST00000375445.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
ENST00000375445.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
ENST00000375445.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
ENST00000375445.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
ENST00000375445.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
ENST00000375445.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
ENST00000375445.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
ENST00000375445.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
ENST00000375445.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
ENST00000419189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
ENST00000419189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
ENST00000419189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
ENST00000419189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
ENST00000419189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
NM_001159736.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
NM_001159736.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
NM_001159736.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
NM_001159736.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
NM_001159736.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
NM_001159736.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
NM_001159736.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
NM_001159736.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
NM_001159736.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
NM_001159736.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
NM_001159736.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
NM_032725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
NM_032725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
NM_032725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
NM_032725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
NM_032725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
NM_032725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
NM_032725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
NM_032725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
NM_032725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
NM_032725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
NM_032725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
NM_032725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012418.hg.1 |
NM_032725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
ENST00000260210.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
ENST00000260210.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
ENST00000260210.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
ENST00000260210.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
ENST00000260210.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
ENST00000260210.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
ENST00000260210.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
ENST00000260210.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
ENST00000260210.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
ENST00000260210.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
ENST00000260210.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
ENST00000260210.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
ENST00000260210.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
ENST00000375445.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
ENST00000375445.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
ENST00000375445.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
ENST00000375445.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
ENST00000375445.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
ENST00000375445.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
ENST00000375445.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
ENST00000375445.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
ENST00000375445.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
ENST00000375445.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
ENST00000375445.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
ENST00000419189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
ENST00000419189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
ENST00000419189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
ENST00000419189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
ENST00000419189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
NM_001159736.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
NM_001159736.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
NM_001159736.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
NM_001159736.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
NM_001159736.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
NM_001159736.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
NM_001159736.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
NM_001159736.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
NM_001159736.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
NM_001159736.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
NM_001159736.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
NM_032725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
NM_032725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
NM_032725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
NM_032725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
NM_032725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
NM_032725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
NM_032725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
NM_032725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
NM_032725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
NM_032725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
NM_032725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
NM_032725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012418.hg.1 |
NM_032725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195304.hg.1 |
ENST00000260210.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195304.hg.1 |
ENST00000375445.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195304.hg.1 |
NM_001159736.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195304.hg.1 |
NM_032725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195304.hg.1 |
ENST00000260210.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195304.hg.1 |
ENST00000375445.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195304.hg.1 |
NM_001159736.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195304.hg.1 |
NM_032725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195304.hg.1 |
TC1100012418.hg.1 |
Clariom D TC |