Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000471966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000471966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000471966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000471966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000471966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000471966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000471966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000487683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000487683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000487683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000487683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000487683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000487683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000487683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000487683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000487683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000487683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000487683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000487683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000487683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000487683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000487683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
ENST00000487683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010853.hg.1 |
TC06000926.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010853.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010853.hg.1 |
ENST00000471966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010853.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600163502.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163502.hg.1 |
ENST00000471966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163502.hg.1 |
ENST00000487683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163502.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600163502.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163502.hg.1 |
ENST00000471966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163502.hg.1 |
ENST00000487683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163502.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600163502.hg.1 |
TC0600009243.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000332958.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000471966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000471966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000471966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000471966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000471966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000471966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000471966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000487683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000487683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000487683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000487683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000487683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000487683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000487683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000487683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000487683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000487683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000487683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000487683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000487683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000487683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000487683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
ENST00000487683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009243.hg.1 |
NM_173560.3 |
Refseq |