Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000295055.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000295055.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000295055.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000295055.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000295055.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000295055.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000295055.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000295055.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000295055.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000295055.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000295055.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000295055.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000295055.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000295055.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000295055.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000295055.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000295055.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000295055.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000295055.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000295055.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000295055.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000295055.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000295055.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000295055.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000295055.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000295055.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000295055.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000295055.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000295055.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000450650.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000450650.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000450650.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000450650.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000450650.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000450650.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000450650.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000450650.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000450650.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000450650.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000450650.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000450650.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000450650.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000450650.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000450650.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000450650.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000458085.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000458085.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000458085.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000458085.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000458085.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000458085.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000458085.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000458085.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000458085.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000458085.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000458085.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000458085.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000458085.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000458085.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000458085.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000458085.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000458085.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000458085.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000458085.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000465450.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000465450.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000465450.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000465450.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000465960.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000465960.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000465960.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000465960.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000465960.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000465960.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000465960.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000465960.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000465960.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000465960.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000465960.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000465960.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000485248.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000485248.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000485248.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000485248.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000485248.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000485248.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000485248.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000490786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000490786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000490786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000490786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
ENST00000490786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
NM_144575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
NM_144575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
NM_144575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
NM_144575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
NM_144575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
NM_144575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
NM_144575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
NM_144575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
NM_144575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
NM_144575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
NM_144575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
NM_144575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
NM_144575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
NM_144575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
NM_144575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
NM_144575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
NM_144575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
NM_144575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
NM_144575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
NM_144575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
NM_144575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
NM_144575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
NM_144575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
NM_144575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
NM_144575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
NM_144575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
NM_144575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
NM_144575.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012159.hg.1 |
NM_144575.2 |
Refseq |