Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004411.hg.1 |
ENST00000283547.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004411.hg.1 |
NM_152324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004411.hg.1 |
TC13000401.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008048.hg.1 |
ENST00000283547.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008048.hg.1 |
ENST00000283547.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008048.hg.1 |
ENST00000283547.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008048.hg.1 |
ENST00000283547.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008048.hg.1 |
ENST00000283547.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008048.hg.1 |
ENST00000283547.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008048.hg.1 |
ENST00000283547.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008048.hg.1 |
ENST00000283547.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008048.hg.1 |
ENST00000497241.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008048.hg.1 |
ENST00000497241.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008048.hg.1 |
ENST00000497241.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008048.hg.1 |
ENST00000497241.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008048.hg.1 |
ENST00000497241.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008048.hg.1 |
ENST00000497241.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008048.hg.1 |
ENST00000497241.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008048.hg.1 |
ENST00000497241.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008048.hg.1 |
ENST00000497241.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008048.hg.1 |
NM_001303133.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008048.hg.1 |
NM_001303133.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008048.hg.1 |
NM_001303133.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008048.hg.1 |
NM_001303133.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008048.hg.1 |
NM_001303133.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008048.hg.1 |
NM_001303133.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008048.hg.1 |
NM_001303133.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008048.hg.1 |
NM_001303133.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008048.hg.1 |
NM_001303133.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008048.hg.1 |
NM_152324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008048.hg.1 |
NM_152324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008048.hg.1 |
NM_152324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008048.hg.1 |
NM_152324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008048.hg.1 |
NM_152324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008048.hg.1 |
NM_152324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008048.hg.1 |
NM_152324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008048.hg.1 |
NM_152324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004412.hg.1 |
ENST00000283547.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004412.hg.1 |
NM_152324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004412.hg.1 |
TC13000401.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1300154177.hg.1 |
ENST00000283547.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154177.hg.1 |
ENST00000497241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154177.hg.1 |
NM_001303133.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154177.hg.1 |
NM_152324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154177.hg.1 |
ENST00000283547.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154177.hg.1 |
ENST00000497241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154177.hg.1 |
NM_001303133.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154177.hg.1 |
NM_152324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154177.hg.1 |
TC1300008048.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300008048.hg.1 |
ENST00000283547.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008048.hg.1 |
ENST00000283547.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008048.hg.1 |
ENST00000283547.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008048.hg.1 |
ENST00000283547.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008048.hg.1 |
ENST00000283547.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008048.hg.1 |
ENST00000283547.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008048.hg.1 |
ENST00000283547.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008048.hg.1 |
ENST00000283547.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008048.hg.1 |
ENST00000497241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008048.hg.1 |
ENST00000497241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008048.hg.1 |
ENST00000497241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008048.hg.1 |
ENST00000497241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008048.hg.1 |
ENST00000497241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008048.hg.1 |
ENST00000497241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008048.hg.1 |
ENST00000497241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008048.hg.1 |
ENST00000497241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008048.hg.1 |
ENST00000497241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008048.hg.1 |
NM_001303133.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008048.hg.1 |
NM_001303133.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008048.hg.1 |
NM_001303133.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008048.hg.1 |
NM_001303133.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008048.hg.1 |
NM_001303133.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008048.hg.1 |
NM_001303133.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008048.hg.1 |
NM_001303133.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008048.hg.1 |
NM_001303133.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008048.hg.1 |
NM_001303133.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008048.hg.1 |
NM_152324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008048.hg.1 |
NM_152324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008048.hg.1 |
NM_152324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008048.hg.1 |
NM_152324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008048.hg.1 |
NM_152324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008048.hg.1 |
NM_152324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008048.hg.1 |
NM_152324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008048.hg.1 |
NM_152324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154176.hg.1 |
ENST00000283547.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154176.hg.1 |
ENST00000497241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154176.hg.1 |
NM_001303133.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154176.hg.1 |
NM_152324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154176.hg.1 |
ENST00000283547.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154176.hg.1 |
ENST00000497241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300154176.hg.1 |
NM_001303133.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154176.hg.1 |
NM_152324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300154176.hg.1 |
TC1300008048.hg.1 |
Clariom D TC |