Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014942.hg.1 |
ENST00000529694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014942.hg.1 |
NM_000890.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014942.hg.1 |
TC11001186.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014949.hg.1 |
ENST00000338350.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014949.hg.1 |
ENST00000529694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014949.hg.1 |
ENST00000533599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014949.hg.1 |
NM_000890.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014949.hg.1 |
TC11001186.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009495.hg.1 |
ENST00000338350.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009495.hg.1 |
ENST00000338350.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009495.hg.1 |
ENST00000338350.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009495.hg.1 |
ENST00000338350.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009495.hg.1 |
ENST00000338350.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009495.hg.1 |
ENST00000338350.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009495.hg.1 |
ENST00000529694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009495.hg.1 |
ENST00000529694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009495.hg.1 |
ENST00000529694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009495.hg.1 |
ENST00000529694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009495.hg.1 |
ENST00000529694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009495.hg.1 |
ENST00000529694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009495.hg.1 |
ENST00000529694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009495.hg.1 |
ENST00000529694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009495.hg.1 |
ENST00000529694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009495.hg.1 |
ENST00000529694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009495.hg.1 |
ENST00000533599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009495.hg.1 |
ENST00000533599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009495.hg.1 |
ENST00000533599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009495.hg.1 |
NM_000890.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009495.hg.1 |
NM_000890.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009495.hg.1 |
NM_000890.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009495.hg.1 |
NM_000890.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009495.hg.1 |
NM_000890.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009495.hg.1 |
NM_000890.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009495.hg.1 |
NM_000890.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014944.hg.1 |
ENST00000338350.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014944.hg.1 |
ENST00000529694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014944.hg.1 |
NM_000890.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014944.hg.1 |
TC11001186.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100171412.hg.1 |
ENST00000338350.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100171412.hg.1 |
ENST00000529694.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100171412.hg.1 |
NM_000890.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100171412.hg.1 |
ENST00000338350.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100171412.hg.1 |
ENST00000529694.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100171412.hg.1 |
NM_000890.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100171412.hg.1 |
TC1100009495.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100171418.hg.1 |
ENST00000338350.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100171418.hg.1 |
ENST00000529694.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100171418.hg.1 |
ENST00000533599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100171418.hg.1 |
NM_000890.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100171418.hg.1 |
ENST00000338350.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100171418.hg.1 |
ENST00000529694.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100171418.hg.1 |
ENST00000533599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100171418.hg.1 |
NM_000890.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100171418.hg.1 |
TC1100009495.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100009495.hg.1 |
ENST00000338350.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009495.hg.1 |
ENST00000338350.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009495.hg.1 |
ENST00000338350.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009495.hg.1 |
ENST00000338350.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009495.hg.1 |
ENST00000338350.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009495.hg.1 |
ENST00000338350.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009495.hg.1 |
ENST00000529694.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009495.hg.1 |
ENST00000529694.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009495.hg.1 |
ENST00000529694.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009495.hg.1 |
ENST00000529694.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009495.hg.1 |
ENST00000529694.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009495.hg.1 |
ENST00000529694.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009495.hg.1 |
ENST00000529694.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009495.hg.1 |
ENST00000529694.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009495.hg.1 |
ENST00000529694.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009495.hg.1 |
ENST00000529694.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009495.hg.1 |
ENST00000533599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009495.hg.1 |
ENST00000533599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009495.hg.1 |
ENST00000533599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009495.hg.1 |
NM_000890.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009495.hg.1 |
NM_000890.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009495.hg.1 |
NM_000890.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009495.hg.1 |
NM_000890.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009495.hg.1 |
NM_000890.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009495.hg.1 |
NM_000890.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009495.hg.1 |
NM_000890.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100171410.hg.1 |
ENST00000529694.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100171410.hg.1 |
NM_000890.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100171410.hg.1 |
ENST00000529694.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100171410.hg.1 |
NM_000890.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100171410.hg.1 |
TC1100009495.hg.1 |
Clariom D TC |