Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07029276.hg.1 |
TC07001885.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07029276.hg.1 |
ENST00000344400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07029276.hg.1 |
ENST00000354475.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07029276.hg.1 |
ENST00000423565.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07029276.hg.1 |
ENST00000485942.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07029276.hg.1 |
NM_014149.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07029275.hg.1 |
TC07001885.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07029275.hg.1 |
ENST00000344400.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07029275.hg.1 |
ENST00000354475.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07029275.hg.1 |
ENST00000423565.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07029275.hg.1 |
NM_014149.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700191489.hg.1 |
ENST00000354475.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700191489.hg.1 |
ENST00000423565.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700191489.hg.1 |
ENST00000466182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700191489.hg.1 |
ENST00000476775.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700191489.hg.1 |
ENST00000479698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700191489.hg.1 |
ENST00000485942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700191489.hg.1 |
NM_014149.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700191489.hg.1 |
ENST00000354475.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700191489.hg.1 |
ENST00000423565.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700191489.hg.1 |
ENST00000466182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700191489.hg.1 |
ENST00000476775.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700191489.hg.1 |
ENST00000479698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700191489.hg.1 |
ENST00000485942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700191489.hg.1 |
NM_014149.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700191489.hg.1 |
TC0700012696.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700191488.hg.1 |
ENST00000354475.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700191488.hg.1 |
ENST00000423565.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700191488.hg.1 |
ENST00000466182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700191488.hg.1 |
ENST00000476775.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700191488.hg.1 |
ENST00000479698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700191488.hg.1 |
NM_014149.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700191488.hg.1 |
ENST00000354475.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700191488.hg.1 |
ENST00000423565.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700191488.hg.1 |
ENST00000466182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700191488.hg.1 |
ENST00000476775.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700191488.hg.1 |
ENST00000479698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700191488.hg.1 |
NM_014149.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700191488.hg.1 |
TC0700012696.hg.1 |
Clariom D TC |