Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000241502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000241502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000241502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000241502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000241502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000241502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000241502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000241502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000241502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000241502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000241502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000241502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000241502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000241502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000241502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000241502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000241502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000241502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000241502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000241502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000241502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000241502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000412924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000412924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000412924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000412924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000412924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000412924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000412924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000412924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000412924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000412924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000412924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000412924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000415708.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000415708.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000415708.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000415708.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000415708.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000415708.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000415708.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000415708.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000415708.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000415708.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000415708.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000415708.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000415708.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000415708.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000415708.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000418169.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000418169.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000418169.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000418169.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000418169.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000418169.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000418169.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000424384.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000424384.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000424384.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000424384.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000424384.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000424384.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000424384.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000424384.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000424384.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000424384.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000424384.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000424384.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000424384.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000424384.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000424384.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000426031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000426031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000426031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000426031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000428738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000428738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000428738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000428738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000428738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000428738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000428738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000428738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000428738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000492360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000492360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000492360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000492360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000494309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000494309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000494309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000494309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000494309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000494309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
ENST00000494309.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_001011537.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_001011537.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_001011537.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_001011537.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_001011537.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_001011537.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_001011537.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_001011537.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_001011537.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_001011537.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_001011537.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_001011537.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_001011537.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_001011537.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_001011537.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_001011537.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_001011537.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_001011537.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_001011537.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_001011537.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_032288.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_032288.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_032288.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_032288.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_032288.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_032288.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_032288.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_032288.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_032288.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_032288.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_032288.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_032288.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_032288.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_032288.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_032288.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_032288.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_032288.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_032288.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_032288.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_032288.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NM_032288.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NR_027840.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NR_027840.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NR_027840.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NR_027840.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NR_027840.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NR_027840.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NR_027840.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NR_027840.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NR_027840.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NR_027840.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NR_027840.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NR_027840.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NR_027840.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NR_027840.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NR_027840.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NR_027840.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NR_027840.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010066.hg.1 |
NR_027840.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011030.hg.1 |
ENST00000424250.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011030.hg.1 |
ENST00000424250.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011030.hg.1 |
ENST00000424250.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011030.hg.1 |
ENST00000424250.2 |
Ensembl |