Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014495.hg.1 |
ENST00000280665.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014495.hg.1 |
ENST00000397173.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014495.hg.1 |
NM_152640.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014495.hg.1 |
TC12001094.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000280665.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000280665.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000280665.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000280665.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000280665.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000280665.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000280665.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000280665.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000280665.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000280665.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000280665.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000280665.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000280665.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000280665.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000280665.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000535873.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000535873.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000535873.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000535873.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000535873.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000535873.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000536665.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000536665.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000536665.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000536665.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000537725.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000537725.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000537725.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000537725.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000537725.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000537725.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000540622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000540622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000540622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000540622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000540622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000540622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000541700.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000541700.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000541700.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000541700.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000541700.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000541700.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000543381.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000543381.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000543381.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000543381.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000543381.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000543381.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000543381.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000543381.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000543381.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000543381.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000543381.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000543381.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
ENST00000543381.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
NM_152640.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
NM_152640.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
NM_152640.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
NM_152640.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
NM_152640.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
NM_152640.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
NM_152640.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
NM_152640.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
NM_152640.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
NM_152640.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
NM_152640.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
NM_152640.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
NM_152640.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
NM_152640.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009590.hg.1 |
NM_152640.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014497.hg.1 |
ENST00000280665.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014497.hg.1 |
ENST00000397173.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014497.hg.1 |
ENST00000540622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014497.hg.1 |
NM_152640.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014497.hg.1 |
TC12001094.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000280665.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000280665.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000280665.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000280665.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000280665.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000280665.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000280665.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000280665.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000280665.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000280665.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000280665.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000280665.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000280665.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000280665.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000280665.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000535873.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000535873.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000535873.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000535873.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000535873.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000535873.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000536665.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000536665.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000536665.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000536665.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000537725.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000537725.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000537725.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000537725.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000537725.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000537725.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000540622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000540622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000540622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000540622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000540622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000540622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000541700.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000541700.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000541700.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000541700.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000541700.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000541700.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000543381.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000543381.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000543381.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000543381.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000543381.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000543381.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000543381.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000543381.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000543381.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000543381.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000543381.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000543381.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
ENST00000543381.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
NM_152640.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
NM_152640.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
NM_152640.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
NM_152640.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
NM_152640.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
NM_152640.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
NM_152640.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
NM_152640.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
NM_152640.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
NM_152640.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
NM_152640.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
NM_152640.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
NM_152640.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
NM_152640.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009590.hg.1 |
NM_152640.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200171753.hg.1 |
ENST00000280665.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171753.hg.1 |
ENST00000540622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171753.hg.1 |
ENST00000543381.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171753.hg.1 |
NM_152640.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200171753.hg.1 |
ENST00000280665.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171753.hg.1 |
ENST00000540622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171753.hg.1 |
ENST00000543381.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171753.hg.1 |
NM_152640.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200171753.hg.1 |
TC1200009590.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200171751.hg.1 |
ENST00000280665.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171751.hg.1 |
ENST00000543381.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171751.hg.1 |
NM_152640.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200171751.hg.1 |
ENST00000280665.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171751.hg.1 |
ENST00000543381.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171751.hg.1 |
NM_152640.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200171751.hg.1 |
TC1200009590.hg.1 |
Clariom D TC |