Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000025399.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000025399.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000025399.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000025399.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000025399.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000025399.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000025399.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000025399.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000025399.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000025399.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000025399.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000025399.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000025399.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000025399.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000025399.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000025399.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000419869.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000419869.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000419869.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000419869.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000419869.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000419869.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000419869.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000419869.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000419869.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000419869.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000419869.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000419869.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000419869.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000419869.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000419869.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000419869.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000419869.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000419869.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000536737.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000536737.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000536737.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000536737.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000538718.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000538718.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000538718.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000538718.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000538718.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000539887.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000539887.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000539887.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000539887.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000539887.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000539887.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000539887.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000541731.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000541731.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000541731.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000541731.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000541731.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000541731.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000541731.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000541731.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000541731.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000541731.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000541731.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000541731.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000541731.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000541731.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000541731.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
ENST00000541731.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
NM_007178.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
NM_007178.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
NM_007178.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
NM_007178.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
NM_007178.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
NM_007178.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
NM_007178.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
NM_007178.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
NM_007178.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
NM_007178.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
NM_007178.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
NM_007178.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
NM_007178.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
NM_007178.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
NM_007178.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
NM_007178.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
NM_007178.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
NM_007178.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006966.hg.1 |
NM_007178.3 |
Refseq |