Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012216.hg.1 |
TC09000918.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012216.hg.1 |
ENST00000380916.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012216.hg.1 |
NM_178566.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009579.hg.1 |
ENST00000380916.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009579.hg.1 |
ENST00000380916.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009579.hg.1 |
ENST00000380916.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009579.hg.1 |
ENST00000380916.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009579.hg.1 |
ENST00000380916.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009579.hg.1 |
ENST00000380916.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009579.hg.1 |
ENST00000380916.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009579.hg.1 |
ENST00000380916.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009579.hg.1 |
ENST00000380916.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009579.hg.1 |
ENST00000380916.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009579.hg.1 |
ENST00000380916.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009579.hg.1 |
ENST00000380916.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009579.hg.1 |
ENST00000380916.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009579.hg.1 |
ENST00000380916.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009579.hg.1 |
ENST00000380916.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009579.hg.1 |
ENST00000497966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009579.hg.1 |
ENST00000497966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009579.hg.1 |
ENST00000497966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009579.hg.1 |
ENST00000497966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009579.hg.1 |
ENST00000497966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009579.hg.1 |
NM_178566.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009579.hg.1 |
NM_178566.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009579.hg.1 |
NM_178566.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009579.hg.1 |
NM_178566.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009579.hg.1 |
NM_178566.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009579.hg.1 |
NM_178566.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009579.hg.1 |
NM_178566.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009579.hg.1 |
NM_178566.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009579.hg.1 |
NM_178566.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009579.hg.1 |
NM_178566.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009579.hg.1 |
NM_178566.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009579.hg.1 |
NM_178566.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009579.hg.1 |
NM_178566.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009579.hg.1 |
NM_178566.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009579.hg.1 |
NM_178566.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009579.hg.1 |
ENST00000380916.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009579.hg.1 |
ENST00000380916.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009579.hg.1 |
ENST00000380916.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009579.hg.1 |
ENST00000380916.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009579.hg.1 |
ENST00000380916.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009579.hg.1 |
ENST00000380916.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009579.hg.1 |
ENST00000380916.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009579.hg.1 |
ENST00000380916.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009579.hg.1 |
ENST00000380916.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009579.hg.1 |
ENST00000380916.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009579.hg.1 |
ENST00000380916.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009579.hg.1 |
ENST00000380916.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009579.hg.1 |
ENST00000380916.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009579.hg.1 |
ENST00000380916.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009579.hg.1 |
ENST00000380916.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009579.hg.1 |
ENST00000497966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009579.hg.1 |
ENST00000497966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009579.hg.1 |
ENST00000497966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009579.hg.1 |
ENST00000497966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009579.hg.1 |
ENST00000497966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009579.hg.1 |
NM_178566.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009579.hg.1 |
NM_178566.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009579.hg.1 |
NM_178566.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009579.hg.1 |
NM_178566.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009579.hg.1 |
NM_178566.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009579.hg.1 |
NM_178566.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009579.hg.1 |
NM_178566.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009579.hg.1 |
NM_178566.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009579.hg.1 |
NM_178566.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009579.hg.1 |
NM_178566.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009579.hg.1 |
NM_178566.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009579.hg.1 |
NM_178566.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009579.hg.1 |
NM_178566.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009579.hg.1 |
NM_178566.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009579.hg.1 |
NM_178566.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165611.hg.1 |
ENST00000380916.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165611.hg.1 |
NM_178566.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165611.hg.1 |
ENST00000380916.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165611.hg.1 |
NM_178566.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165611.hg.1 |
TC0900009579.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900165610.hg.1 |
ENST00000380916.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165610.hg.1 |
NM_178566.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165610.hg.1 |
ENST00000380916.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900165610.hg.1 |
NM_178566.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900165610.hg.1 |
TC0900009579.hg.1 |
Clariom D TC |