Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002660.hg.1 |
TC14002295.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002660.hg.1 |
ENST00000280082.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002660.hg.1 |
ENST00000556784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002660.hg.1 |
NM_054024.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
ENST00000280082.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
ENST00000280082.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
ENST00000280082.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
ENST00000280082.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
ENST00000280082.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
ENST00000280082.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
ENST00000280082.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
ENST00000280082.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
ENST00000280082.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
ENST00000280082.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
ENST00000280082.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
ENST00000280082.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
ENST00000280082.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
ENST00000555143.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
ENST00000555143.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
ENST00000555143.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
ENST00000555143.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
ENST00000555143.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
ENST00000555143.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
ENST00000556784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
ENST00000556784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
ENST00000556784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
ENST00000556784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
ENST00000556784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
ENST00000556784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
ENST00000557148.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
ENST00000557148.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
ENST00000557148.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
ENST00000557148.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
ENST00000557148.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
ENST00000557148.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
ENST00000557148.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
NM_054024.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
NM_054024.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
NM_054024.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
NM_054024.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
NM_054024.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
NM_054024.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
NM_054024.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
NM_054024.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
NM_054024.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
NM_054024.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
NM_054024.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010604.hg.1 |
NM_054024.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400179294.hg.1 |
ENST00000280082.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400179294.hg.1 |
ENST00000556784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400179294.hg.1 |
ENST00000557148.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400179294.hg.1 |
NM_054024.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400179294.hg.1 |
ENST00000280082.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400179294.hg.1 |
ENST00000556784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400179294.hg.1 |
ENST00000557148.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400179294.hg.1 |
NM_054024.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400179294.hg.1 |
TC1400010604.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400179295.hg.1 |
ENST00000280082.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400179295.hg.1 |
ENST00000556784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400179295.hg.1 |
NM_054024.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400179295.hg.1 |
ENST00000280082.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400179295.hg.1 |
ENST00000556784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400179295.hg.1 |
NM_054024.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400179295.hg.1 |
TC1400010604.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
ENST00000280082.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
ENST00000280082.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
ENST00000280082.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
ENST00000280082.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
ENST00000280082.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
ENST00000280082.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
ENST00000280082.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
ENST00000280082.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
ENST00000280082.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
ENST00000280082.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
ENST00000280082.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
ENST00000280082.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
ENST00000280082.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
ENST00000555143.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
ENST00000555143.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
ENST00000555143.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
ENST00000555143.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
ENST00000555143.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
ENST00000555143.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
ENST00000556784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
ENST00000556784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
ENST00000556784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
ENST00000556784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
ENST00000556784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
ENST00000556784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
ENST00000557148.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
ENST00000557148.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
ENST00000557148.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
ENST00000557148.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
ENST00000557148.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
ENST00000557148.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
ENST00000557148.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
NM_054024.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
NM_054024.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
NM_054024.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
NM_054024.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
NM_054024.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
NM_054024.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
NM_054024.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
NM_054024.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
NM_054024.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
NM_054024.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
NM_054024.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010604.hg.1 |
NM_054024.3 |
Refseq |