Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000379995.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000379995.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000379995.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000379995.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000379995.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000379995.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000379995.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000379997.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000379997.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000379997.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000379997.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000379997.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000379997.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000379997.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000379997.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000380003.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000380003.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000380003.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000380003.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000380003.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000380003.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000380003.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000380003.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000380003.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000380003.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000380003.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000380003.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000380003.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000461679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000461679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000461679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000461679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000461679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000461679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000461679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000461679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000488117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000488117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000488117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000488117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000488117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000488117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000488117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000488117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000488117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000488117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000488117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000488117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000488117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000488117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000619707.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000619707.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000619707.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000619707.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000619707.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000619707.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000619707.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000619707.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000619707.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000619707.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000619707.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000619707.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000619707.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000619707.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000619707.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
ENST00000619707.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
NM_001256054.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
NM_001256054.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
NM_001256054.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
NM_001256054.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
NM_001256054.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
NM_001256054.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
NM_001256054.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
NM_001256054.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
NM_001256054.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
NM_001256054.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
NM_001256054.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
NM_001256054.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
NM_001256054.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
NM_001256054.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
NM_001256054.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
NM_001256054.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
NM_018325.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
NM_018325.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
NM_018325.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
NM_018325.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
NM_018325.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
NM_018325.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
NM_018325.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
NM_018325.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
NM_018325.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
NM_018325.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
NM_018325.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
NM_018325.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
NM_018325.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
NM_018325.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
NM_145005.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
NM_145005.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
NM_145005.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
NM_145005.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
NM_145005.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
NM_145005.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
NM_145005.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
NM_145005.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009779.hg.1 |
NM_145005.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013000.hg.1 |
TC09000990.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013000.hg.1 |
ENST00000380003.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013000.hg.1 |
ENST00000488117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013000.hg.1 |
NM_001256054.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013000.hg.1 |
NM_018325.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166923.hg.1 |
ENST00000380003.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166923.hg.1 |
ENST00000488117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166923.hg.1 |
ENST00000619707.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166923.hg.1 |
NM_001256054.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166923.hg.1 |
NM_018325.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166923.hg.1 |
ENST00000380003.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166923.hg.1 |
ENST00000488117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166923.hg.1 |
ENST00000619707.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166923.hg.1 |
NM_001256054.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166923.hg.1 |
NM_018325.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166923.hg.1 |
TC0900009779.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000379995.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000379995.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000379995.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000379995.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000379995.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000379995.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000379995.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000379997.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000379997.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000379997.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000379997.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000379997.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000379997.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000379997.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000379997.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000380003.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000380003.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000380003.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000380003.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000380003.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000380003.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000380003.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000380003.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000380003.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000380003.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000380003.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000380003.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000380003.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000461679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000461679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000461679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000461679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000461679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000461679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000461679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000461679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000488117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000488117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000488117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000488117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000488117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000488117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000488117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000488117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000488117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000488117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000488117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000488117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000488117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000488117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000619707.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000619707.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000619707.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000619707.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000619707.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000619707.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000619707.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000619707.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000619707.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000619707.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000619707.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000619707.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000619707.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000619707.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000619707.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
ENST00000619707.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
NM_001256054.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
NM_001256054.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
NM_001256054.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
NM_001256054.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
NM_001256054.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
NM_001256054.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
NM_001256054.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
NM_001256054.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
NM_001256054.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
NM_001256054.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
NM_001256054.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
NM_001256054.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
NM_001256054.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
NM_001256054.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
NM_001256054.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
NM_001256054.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
NM_018325.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
NM_018325.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
NM_018325.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
NM_018325.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
NM_018325.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
NM_018325.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
NM_018325.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
NM_018325.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
NM_018325.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
NM_018325.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
NM_018325.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
NM_018325.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
NM_018325.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
NM_018325.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
NM_145005.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
NM_145005.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
NM_145005.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
NM_145005.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
NM_145005.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
NM_145005.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
NM_145005.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
NM_145005.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009779.hg.1 |
NM_145005.6 |
Refseq |