Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNRC6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015088.2 | NP_055903.2 | 5 - 6 | 3014 | 58 | |
GenBank mRNA | AB029016.2 | - | 5 - 6 | 2997 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Homo sapiens Probe spans exons |