Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001236.hg.1 |
TC03000051.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001236.hg.1 |
ENST00000287647.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001236.hg.1 |
ENST00000383806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001236.hg.1 |
ENST00000383807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001236.hg.1 |
ENST00000419585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001236.hg.1 |
NM_001018115.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001236.hg.1 |
NM_033084.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145620.hg.1 |
ENST00000287647.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145620.hg.1 |
ENST00000383807.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145620.hg.1 |
ENST00000419585.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145620.hg.1 |
ENST00000421731.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145620.hg.1 |
ENST00000470028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145620.hg.1 |
NM_001018115.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145620.hg.1 |
NM_033084.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145620.hg.1 |
ENST00000287647.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145620.hg.1 |
ENST00000383807.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145620.hg.1 |
ENST00000419585.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145620.hg.1 |
ENST00000421731.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145620.hg.1 |
ENST00000470028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300145620.hg.1 |
NM_001018115.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145620.hg.1 |
NM_033084.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300145620.hg.1 |
TC0300006575.hg.1 |
Clariom D TC |