Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032135.3 | NP_115511.3 | 1 | 95 | ||
GenBank mRNA | AK124110.1 | - | 2 | 2522 | 95 | |
AK302615.1 | - | 1 | 95 | |||
AL136775.1 | - | 1 | 2216 | 95 | ||
BC039878.1 | - | 1 | 5163160 | 95 | ||
CR933608.1 | - | 1 | 2872 | 95 | ||
CR933626.1 | - | 1 | 2846 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |