Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011625.hg.1 |
TC0X000872.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011625.hg.1 |
ENST00000324138.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011625.hg.1 |
ENST00000398334.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011625.hg.1 |
NM_001018113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011625.hg.1 |
NM_152633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000324138.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000324138.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000324138.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000324138.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000324138.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000324138.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000324138.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000324138.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000324138.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000324138.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000324138.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000324138.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000324138.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000324138.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000398334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000398334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000398334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000398334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000398334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000398334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000398334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000398334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000398334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000398334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000398334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000398334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000398334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000398334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000398334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000452869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000452869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000452869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000452869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000452869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000452869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000452869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000452869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000452869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000452869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000452869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000452869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000489126.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000489126.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000489126.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
ENST00000489126.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
NM_001018113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
NM_001018113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
NM_001018113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
NM_001018113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
NM_001018113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
NM_001018113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
NM_001018113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
NM_001018113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
NM_001018113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
NM_001018113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
NM_001018113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
NM_001018113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
NM_001018113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
NM_001018113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
NM_001018113.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
NM_152633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
NM_152633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
NM_152633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
NM_152633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
NM_152633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
NM_152633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
NM_152633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
NM_152633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
NM_152633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
NM_152633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
NM_152633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
NM_152633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
NM_152633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009117.hg.1 |
NM_152633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000324138.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000324138.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000324138.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000324138.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000324138.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000324138.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000324138.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000324138.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000324138.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000324138.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000324138.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000324138.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000324138.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000324138.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000398334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000398334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000398334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000398334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000398334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000398334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000398334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000398334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000398334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000398334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000398334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000398334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000398334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000398334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000398334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000452869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000452869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000452869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000452869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000452869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000452869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000452869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000452869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000452869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000452869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000452869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000452869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000489126.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000489126.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000489126.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
ENST00000489126.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
NM_001018113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
NM_001018113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
NM_001018113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
NM_001018113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
NM_001018113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
NM_001018113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
NM_001018113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
NM_001018113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
NM_001018113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
NM_001018113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
NM_001018113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
NM_001018113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
NM_001018113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
NM_001018113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
NM_001018113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
NM_152633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
NM_152633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
NM_152633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
NM_152633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
NM_152633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
NM_152633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
NM_152633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
NM_152633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
NM_152633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
NM_152633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
NM_152633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
NM_152633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
NM_152633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009117.hg.1 |
NM_152633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00162476.hg.1 |
ENST00000324138.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00162476.hg.1 |
ENST00000398334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00162476.hg.1 |
ENST00000452869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00162476.hg.1 |
NM_001018113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00162476.hg.1 |
NM_152633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00162476.hg.1 |
ENST00000324138.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00162476.hg.1 |
ENST00000398334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00162476.hg.1 |
ENST00000452869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00162476.hg.1 |
NM_001018113.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00162476.hg.1 |
NM_152633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00162476.hg.1 |
TC0X00009117.hg.1 |
Clariom D TC |