Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000360688.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000360688.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000360688.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000360688.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000360688.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000360688.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000360688.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000360688.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000360688.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000360688.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000360688.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000360688.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000360688.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000360688.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000406256.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000406256.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000406256.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000406256.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000406256.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000406256.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000406256.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000406256.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000406256.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000406256.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000406256.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000406256.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000406256.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000406256.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000565704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000565704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000565704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000565704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000565704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000565704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000565704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000566434.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000566434.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000566434.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000566434.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000566434.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000566434.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000566613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000566613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
ENST00000566613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
NM_024307.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
NM_024307.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
NM_024307.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
NM_024307.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
NM_024307.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
NM_024307.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
NM_024307.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
NM_024307.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
NM_024307.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
NM_024307.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
NM_024307.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
NM_024307.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009953.hg.1 |
NM_024307.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16014585.hg.1 |
ENST00000360688.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16014585.hg.1 |
ENST00000406256.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16014585.hg.1 |
NM_024307.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16014585.hg.1 |
TC16001020.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600176756.hg.1 |
ENST00000360688.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600176756.hg.1 |
ENST00000406256.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600176756.hg.1 |
ENST00000565704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600176756.hg.1 |
ENST00000566434.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600176756.hg.1 |
ENST00000566613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600176756.hg.1 |
NM_024307.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600176756.hg.1 |
ENST00000360688.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600176756.hg.1 |
ENST00000406256.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600176756.hg.1 |
ENST00000565704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600176756.hg.1 |
ENST00000566434.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600176756.hg.1 |
ENST00000566613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600176756.hg.1 |
NM_024307.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600176756.hg.1 |
TC1600009953.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000360688.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000360688.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000360688.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000360688.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000360688.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000360688.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000360688.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000360688.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000360688.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000360688.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000360688.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000360688.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000360688.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000360688.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000406256.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000406256.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000406256.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000406256.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000406256.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000406256.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000406256.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000406256.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000406256.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000406256.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000406256.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000406256.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000406256.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000406256.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000565704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000565704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000565704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000565704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000565704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000565704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000565704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000566434.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000566434.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000566434.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000566434.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000566434.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000566434.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000566613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000566613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
ENST00000566613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
NM_024307.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
NM_024307.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
NM_024307.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
NM_024307.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
NM_024307.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
NM_024307.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
NM_024307.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
NM_024307.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
NM_024307.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
NM_024307.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
NM_024307.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
NM_024307.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009953.hg.1 |
NM_024307.2 |
Refseq |